APO455825896 Apollo Scientific Ltd

5-Methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD00090754
  • Cas Number : 402-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236-240
Synonym 3-Carboxy-5-methyl-1H-pyrazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-methyl-1h-pyrazole-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/5-methyl-1h-pyrazole-3-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart