APO455825905 Apollo Scientific Ltd

Imidazo[2,1-b][1,3]thiazole-6-carboxylic acid

  • MDL number MFCD03419463
  • Cas Number : 53572-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Carboxyimidazo[2,1-b][1,3]thiazole
Storage temp. Store at 2-8
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29047

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=29047
Availability details for
Quantity
  • Availability
  • Add to Cart