APO455825911 Apollo Scientific Ltd

(4-Phenyl-1,3-thiazol-2-yl)acetonitrile

  • MDL number MFCD00551791
  • Cas Number : 41381-89-9
  • Molecular Weight : 200.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-60
Synonym 2-(Cyanomethyl)-4-phenyl-1,3-thiazole, [2-(Cyanomethyl)-1,3-thiazol-4-yl]benzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-phenyl-13-thiazol-2-ylacetonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-phenyl-13-thiazol-2-ylacetonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart