APO455825918 Apollo Scientific Ltd

4-(Piperidin-1-yl)benzoic acid

  • MDL number MFCD00587602
  • Cas Number : 22090-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214-219
Synonym 1-(4-Carboxyphenyl)piperidine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-piperidin-1-ylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-piperidin-1-ylbenzoic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart