APO455825936 Apollo Scientific Ltd

1-(2-Aminoethyl)-4-methylpiperazine

  • MDL number MFCD03701701
  • Cas Number : 934-98-5
  • Molecular Weight : 143.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(4-Methylpiperazin-1-yl)ethylamine, 1-Amino-2-(4-methylpiperazin-1-yl)ethane
Purity 95
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 112/30mm
Product Link https://store.apolloscientific.co.uk/product/1-2-aminoethyl-4-methylpiperazine

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-2-aminoethyl-4-methylpiperazine
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart