APO455825937 Apollo Scientific Ltd

3-Amino-1-methylpiperidine dihydrochloride

  • MDL number MFCD06796280
  • Cas Number : 894808-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236-238
Synonym 1-Methylpiperidin-3-amine dihydrochloride
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28467

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28467
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart