APO455825940 Apollo Scientific Ltd

4-[2-(Pyrrolidin-1-yl)ethyl]piperidine

  • MDL number MFCD03092939
  • Cas Number : 14759-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[2-(Piperidin-4-yl)ethyl]pyrrolidine
Purity 98
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 120/4mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28435

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28435
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart