APO455825942 Apollo Scientific Ltd

1-Carbamoylpiperidine-4-carboxylic acid

  • MDL number MFCD06655891
  • Cas Number : 851168-77-9
  • Molecular Weight : 172.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196(dec.)
Synonym 1-Carbamoyl-4-carboxypiperidine, 1-(Aminocarbonyl)-4-carboxypiperidine, 4-Carboxypiperidine-1-carboxamide
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-carbamoylpiperidine-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-carbamoylpiperidine-4-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart