APO455825956 Apollo Scientific Ltd

4-(Aminomethyl)benzaldehyde, N-BOC protected

  • MDL number MFCD06659090
  • Cas Number : 156866-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82.9-83.7
Synonym tert-Butyl (4-formylbenzyl)carbamate, 4-{[(tert-Butoxycarbonyl)amino]methyl}benzaldehyde
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-aminomethylbenzaldehyde-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-aminomethylbenzaldehyde-n-boc-protected
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart