APO455825959 Apollo Scientific Ltd

2-(Piperazin-1-yl)-1,3,5-triazine

  • MDL number MFCD04973348
  • Cas Number : 59215-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-79
Synonym 1-(1,3,5-Triazin-2-yl)piperazine, 2-(Piperazin-1-yl)-s-triazine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28477

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28477
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart