APO455825961 Apollo Scientific Ltd

1-(Methylsulphonyl)piperazine

  • MDL number MFCD02153225
  • Cas Number : 55276-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-88
Synonym Methyl piperazin-1-yl sulphone
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1-methylsulphonylpiperazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-methylsulphonylpiperazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart