APO455825979 Apollo Scientific Ltd

1-Cyclohexylpiperazine

  • MDL number MFCD00044809
  • Cas Number : 17766-28-8
  • Molecular Weight : 168.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 32-34
Iupac Name 1-cyclohexylpiperazine
Inchi Code 1S/C10H20N2/c1-2-4-10(5-3-1)12-8-6-11-7-9-12/h10-11H,1-9H2
Inchi Key XPDSXKIDJNKIQY-UHFFFAOYSA-N

Description

Synonym (Piperazin-1-yl)cyclohexane
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 32-34
Product Link https://store.apolloscientific.co.uk/product/1-cyclohexylpiperazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-cyclohexylpiperazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart