APO455825981 Apollo Scientific Ltd

Ethyl 1,4-dihydro-2-(methylthio)-4-oxopyrimidine-5-carboxylate

  • MDL number MFCD03002357
  • Cas Number : 53554-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-137
Synonym Ethyl 1,4-dihydro-2-(methylsulphanyl)-4-oxopyrimidine-5-carboxylate, 1,4-Dihydro-5-(ethoxycarbonyl)-2-(methylthio)-4-oxopyrimidine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-14-dihydro-2-methylthio-4-oxopyrimidine-5-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-14-dihydro-2-methylthio-4-oxopyrimidine-5-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart