APO455825982 Apollo Scientific Ltd

4-(Dimethylamino)piperidine

  • MDL number MFCD00023144
  • Cas Number : 50533-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N,N-Dimethylpiperidin-4-amine
Density 1.09
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 187
Product Link https://store.apolloscientific.co.uk/product/4-dimethylaminopiperidine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-dimethylaminopiperidine
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart