APO455825985 Apollo Scientific Ltd

Imidazo[2,1-b][1,3]benzothiazole-2-carboxylic acid hydrate

  • MDL number MFCD08444056
  • Cas Number : 1284226-74-9
  • Molecular Weight : 236.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 266-268
Synonym 2-Carboxyimidazo[2,1-b][1,3]benzothiazole hydrate
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/imidazo21-b13benzothiazole-2-carboxylic-acid-hydrate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/imidazo21-b13benzothiazole-2-carboxylic-acid-hydrate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart