APO455825987 Apollo Scientific Ltd

1-Methyl-4-(piperidin-4-yl)piperazine

  • MDL number MFCD03274729
  • Cas Number : 53617-36-0
  • Molecular Weight : 183.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-56
Synonym 4-(4-Methylpiperazin-1-yl)piperidine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-methyl-4-piperidin-4-ylpiperazine

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-methyl-4-piperidin-4-ylpiperazine
pictograms GHS05;GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart