APO455825989 Apollo Scientific Ltd

1-(4-Methylpyridin-2-yl)piperazine

  • MDL number MFCD03412118
  • Cas Number : 34803-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Methyl-2-(piperazin-1-yl)pyridine
Purity 97
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 161-163/8mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28511

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28511
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart