APO455826020 Apollo Scientific Ltd

1-(Piperidin-1-ylsulphonyl)piperazine

  • MDL number MFCD04973338
  • Cas Number : 500587-48-4
  • Molecular Weight : 233.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-piperidin-1-ylsulphonylpiperazine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-piperidin-1-ylsulphonylpiperazine
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart