APO455826021 Apollo Scientific Ltd

1-(2-Propoxyethyl)piperazine

  • MDL number MFCD04117823
  • Cas Number : 741667-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-{[2-(Prop-1-yl)oxy]ethyl}piperazine
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 107/12mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28532

Safety Information

signalword Warning
pcodes P271;P280;P260
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28532
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart