APO455826023 Apollo Scientific Ltd

1-(1,3-Thiazol-2-yl)piperazine

  • MDL number MFCD00778419
  • Cas Number : 42270-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1-13-thiazol-2-ylpiperazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-13-thiazol-2-ylpiperazine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart