APO455826031 Apollo Scientific Ltd

4-Amino-5-(thien-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00269270
  • Cas Number : 61019-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219-220
Synonym 4-Amino-3-mercapto-5-(thien-2-yl)-4H-1,2,4-triazole, 4-Amino-2,4-dihydro-5-(thien-2-yl)-3H-1,2,4-triazole-3-thione
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12682

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12682
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart