APO455826058 Apollo Scientific Ltd

1,8-Naphthyridine-2-carboxylic acid

  • MDL number MFCD00202900
  • Cas Number : 215523-34-5
  • Molecular Weight : 174.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220(dec.)
Synonym 2-Carboxy-1,8-naphthyridine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/18-naphthyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/18-naphthyridine-2-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart