APO455826074 Apollo Scientific Ltd

1-Bromo-3-chloro-5,5-dimethylhydantoin

  • MDL number MFCD00037350
  • Cas Number : 16079-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160.2-161.5
Synonym 1-Bromo-3-chloro-5,5-dimethylimidazolidine-2,4-dione, 1-Bromo-3-chloro-5,5-dimethyl-2,4-dioxoimidazolidine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16287

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16287
pictograms GHS03;GHS05;GHS07
hstatements H272;H314;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart