APO455826109 Apollo Scientific Ltd

5-Chloro-2,3,3-trimethyl-3H-indole

  • MDL number MFCD00126966
  • Cas Number : 25981-83-3
  • Molecular Weight : 193.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Chloro-2,3,3-trimethylindolenine
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-233-trimethyl-3h-indole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-233-trimethyl-3h-indole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart