APO455826110 Apollo Scientific Ltd

7-Hydroxy-1H-indole

  • MDL number MFCD00152102
  • Cas Number : 2380-84-9
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-88
Synonym 1H-Indol-7-ol
Purity 97
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-hydroxy-1h-indole

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/7-hydroxy-1h-indole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart