APO455826116 Apollo Scientific Ltd

4-(Methylamino)tetrahydro-2H-pyran

  • MDL number MFCD06658492
  • Cas Number : 220641-87-2
  • Molecular Weight : 115.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Methyltetrahydro-2H-pyran-4-amine, 4-(Methylamino)oxane
Purity 95
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methylaminotetrahydro-2h-pyran

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylaminotetrahydro-2h-pyran
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart