APO455826122 Apollo Scientific Ltd

Dibenzo[b,d]furan-2-sulphonyl chloride

  • MDL number MFCD00094222
  • Cas Number : 23602-98-4
  • Molecular Weight : 266.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name dibenzo[b,d]furan-2-sulfonyl chloride
Inchi Code 1S/C12H7ClO3S/c13-17(14,15)8-5-6-12-10(7-8)9-3-1-2-4-11(9)16-12/h1-7H
Inchi Key ULCKEERECJMLHP-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/dibenzobdfuran-2-sulphonyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/dibenzobdfuran-2-sulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart