APO455826128 Apollo Scientific Ltd

5-Aminobenzo[b]thiophene-2-carboxylic acid

  • MDL number MFCD01159705
  • Cas Number : 98589-46-9
  • Molecular Weight : 193.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 274-278
Synonym 5-Amino-2-carboxybenzo[b]thiophene, 5-Amino-2-carboxy-1-benzothiophene
Purity 95
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-aminobenzobthiophene-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-aminobenzobthiophene-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart