APO455826140 Apollo Scientific Ltd

5-Amino-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD00973833
  • Cas Number : 4058-91-7
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Amino-4-carboxy-1-methyl-1H-pyrazole
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-1-methyl-1h-pyrazole-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-1-methyl-1h-pyrazole-4-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart