APO455826145 Apollo Scientific Ltd

4-(Aminomethyl)tetrahydro-2H-pyran

  • MDL number MFCD02179435
  • Cas Number : 130290-79-8
  • Molecular Weight : 115.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (Tetrahydro-2H-pyran-4-yl)methylamine, 4-(Aminomethyl)oxane
Purity 98
Density 1.02
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 75-76/18mm
Product Link https://store.apolloscientific.co.uk/product/4-aminomethyltetrahydro-2h-pyran

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-aminomethyltetrahydro-2h-pyran
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart