APO455826146 Apollo Scientific Ltd

5-Amino-2-oxindole

  • MDL number MFCD02179603
  • Cas Number : 20876-36-2
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-186
Synonym 5-Amino-1,3-dihydro-2H-indol-2-one, 5-Aminoindolin-2-one
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-2-oxindole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-2-oxindole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart