APO455826153 Apollo Scientific Ltd

4-Aminotetrahydro-2H-pyran acetate

  • MDL number MFCD08445669
  • Cas Number : 1005498-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-140
Synonym Tetrahydro-2H-pyran-4-amine acetate, 4-Aminooxane acetate
Purity 98
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31364

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31364
pictograms GHS02;GHS07
hstatements H228;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart