APO455826154 Apollo Scientific Ltd

4-Aminotetrahydro-2H-pyran hydrochloride

  • MDL number MFCD00100881
  • Cas Number : 33024-60-1
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230-250
Synonym Tetrahydro-2H-pyran-4-amine hydrochloride, 4-Aminooxane hydrochloride, Oxan-4-amine hydrochloride
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-aminotetrahydro-2h-pyran-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-aminotetrahydro-2h-pyran-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart