APO455826157 Apollo Scientific Ltd

3-Benzoyl-1-[(4-methylphenyl)sulphonyl]pyrrole

  • MDL number MFCD03411960
  • Cas Number : 139261-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-96
Synonym 3-Benzoyl-1-tosylpyrrole
Purity 96
Storage temp. Ambient temperature
Physical Form solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16499

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16499
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart