APO455826160 Apollo Scientific Ltd

4-(Benzylcarbamoyl)benzeneboronic acid

  • MDL number MFCD03411953
  • Cas Number : 252663-47-1
  • Molecular Weight : 255.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212-218
Synonym 4-[(Benzylamino)carbonyl]benzeneboronic acid, N-Benzyl-4-boronobenzamide
Purity 98
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-benzylcarbamoylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-benzylcarbamoylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart