APO455826166 Apollo Scientific Ltd

4-(2-Aminopyrimidin-4-yl)benzoic acid

  • MDL number MFCD03407445
  • Cas Number : 216959-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-4-(4-carboxyphenyl)pyrimidine, 2-Amino-4-(4-carboxyphenyl)-1,3-diazine
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31425

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31425
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart