APO455826170 Apollo Scientific Ltd

7-Aminopyrazolo[1,5-a]pyrimidine-6-carboxylic acid

  • MDL number MFCD00973861
  • Cas Number : 68262-33-9
  • Molecular Weight : 178.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242
Synonym 7-Amino-6-carboxypyrazolo[1,5-a]pyrimidine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-aminopyrazolo15-apyrimidine-6-carboxylic-acid

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/7-aminopyrazolo15-apyrimidine-6-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart