APO455826171 Apollo Scientific Ltd

2,3-Dihydro-7-nitrobenzo[b]furan-5-sulphonyl chloride

  • MDL number MFCD03408011
  • Cas Number : 874781-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-113
Synonym 5-(Chlorosulphonyl)-2,3-dihydro-7-nitrobenzo[b]furan, 5-(Chlorosulphonyl)-2,3-dihydro-7-nitro-1-benzofuran
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31410

Safety Information

signalword Danger
pcodes P280;P301+P330+P331
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31410
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart