APO455826173 Apollo Scientific Ltd

(1,3-Benzothiazol-2-yl)acetic acid

  • MDL number MFCD00483536
  • Cas Number : 29182-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-113
Synonym 2-(Carboxymethyl)-1,3-benzothiazole, 2-(1,3-Benzothiazol-2-yl)ethanoic acid
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31378

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31378
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart