APO455826176 Apollo Scientific Ltd

2-(Chloromethyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole

  • MDL number MFCD01230030
  • Cas Number : 24023-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-[5-(Chloromethyl)-1,3,4-oxadiazol-2-yl]anisole
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31402

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31402
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart