APO455826182 Apollo Scientific Ltd

4-Bromo-1-trityl-1H-imidazole

  • MDL number MFCD00185657
  • Cas Number : 87941-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242-246
Synonym 4-Bromo-1-[tris(phenyl)methyl]-1H-imidazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 492
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16526

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16526
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart