APO455826188 Apollo Scientific Ltd

7-Bromo-2-oxindole

  • MDL number MFCD02179613
  • Cas Number : 320734-35-8
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 7-Bromo-1,3-dihydro-2H-indol-2-one
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-bromo-2-oxindole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/7-bromo-2-oxindole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart