APO455826189 Apollo Scientific Ltd

4-Bromo-1H-pyrrole-2-carboxaldehyde

  • MDL number MFCD02179586
  • Cas Number : 931-33-9
  • Molecular Weight : 174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-2-formyl-1H-pyrrole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-1h-pyrrole-2-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-1h-pyrrole-2-carboxaldehyde
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart