APO455826191 Apollo Scientific Ltd

3-(tert-Butoxycarbonyl)benzeneboronic acid

  • MDL number MFCD01630855
  • Cas Number : 220210-56-0
  • Molecular Weight : 222.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-102
Synonym 3-(tert-Butoxycarbonyl)phenylboronic acid, tert-Butyl 3-boronobenzoate
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-tert-butoxycarbonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-tert-butoxycarbonylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart