APO455826192 Apollo Scientific Ltd

4-(tert-Butoxycarbonyl)benzeneboronic acid

  • MDL number MFCD03411946
  • Cas Number : 850568-54-6
  • Molecular Weight : 222.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-106
Synonym 4-(tert-Butoxycarbonyl)phenylboronic acid, tert-Butyl 4-boronobenzoate
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-tert-butoxycarbonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-tert-butoxycarbonylbenzeneboronic-acid
pictograms GHS07
hstatements H302;H312;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart