APO455826195 Apollo Scientific Ltd

4-(Butylcarbamoyl)benzeneboronic acid

  • MDL number MFCD03788412
  • Cas Number : 252663-48-2
  • Molecular Weight : 221.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Borono-N-butylbenzamide
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-butylcarbamoylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P260;P280;P301+P310
MSDS https://store.apolloscientific.co.uk/product/4-butylcarbamoylbenzeneboronic-acid
pictograms GHS06;GHS07
hstatements H335;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart