APO455826196 Apollo Scientific Ltd

4-(tert-Butylaminocarbonyl)benzeneboronic acid

  • MDL number MFCD03788413
  • Cas Number : 850568-14-8
  • Molecular Weight : 221.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230-246
Purity 97
Storage temp. Store at 2-8
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-tert-butylaminocarbonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-tert-butylaminocarbonylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart