APO455826199 Apollo Scientific Ltd

4-Carboxy-2-nitrobenzeneboronic acid

  • MDL number MFCD00448780
  • Cas Number : 85107-54-6
  • Molecular Weight : 210.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 265-268
Iupac Name 4-(dihydroxyboryl)-3-nitrobenzoic acid
Inchi Code 1S/C7H6BNO6/c10-7(11)4-1-2-5(8(12)13)6(3-4)9(14)15/h1-3,12-13H,(H,10,11)
Inchi Key APBXPKJLXBBPQJ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 265-268
Product Link https://store.apolloscientific.co.uk/product/4-carboxy-2-nitrobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-carboxy-2-nitrobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart