APO455826200 Apollo Scientific Ltd

2-Oxindole-5-carboxylic acid

  • MDL number MFCD03411961
  • Cas Number : 102359-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 310-316
Synonym 5-Carboxyindolin-2-one, 2,3-Dihydro-2-oxo-1H-indole-5-carboxylic acid
Purity 97
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-oxindole-5-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-oxindole-5-carboxylic-acid
pictograms GHS07
hstatements H302;H317
Availability details for
Quantity
  • Availability
  • Add to Cart