APO455826201 Apollo Scientific Ltd

2-Carboxybenzeneboronic acid

  • MDL number MFCD01318118
  • Cas Number : 149105-19-1
  • Molecular Weight : 165.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220
Synonym (2-Dihydroxyboronyl)benzoic acid, 2-Boronobenzoic acid
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-carboxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340
MSDS https://store.apolloscientific.co.uk/product/2-carboxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart